Anti-LRFN1 Mouse Monoclonal Antibody [clone: 610712]

Supplier: R & D Systems
MAB5669
102886-074EA 584.25 USD
102886-074
Anti-LRFN1 Mouse Monoclonal Antibody [clone: 610712]
Antibodies
Type: Primary
Antigen: LRFN1
Clonality: Monoclonal
Clone: 610712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR