Anti-EPHB3 Mouse Monoclonal Antibody [clone: 647308]

Supplier: R & D Systems
MAB5667
102886-070EA 594.37 USD
102886-070
Anti-EPHB3 Mouse Monoclonal Antibody [clone: 647308]
Antibodies
Type: Primary
Antigen: EPHB3
Clonality: Monoclonal
Clone: 647308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR