Anti-NRP1 Mouse Monoclonal Antibody [clone: 130604]

Supplier: R & D Systems
MAB5661
220021-222EA 763.57 USD
220021-222
Anti-NRP1 Mouse Monoclonal Antibody [clone: 130604]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 130604
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR