Anti-CES2 Mouse Monoclonal Antibody [clone: 720711]

Supplier: R & D Systems
MAB5657
102886-068EA 594.37 USD
102886-068
Anti-CES2 Mouse Monoclonal Antibody [clone: 720711]
Antibodies
Type: Primary
Antigen: CES2
Clonality: Monoclonal
Clone: 720711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR