Anti-IFNG Mouse Monoclonal Antibody [clone: 133816]

Supplier: R & D Systems
MAB565
220021-220EA 772.25 USD
220021-220
Anti-IFNG Mouse Monoclonal Antibody [clone: 133816]
Antibodies
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 133816
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR