Anti-NFATC1 (phospho-Ser172) Mouse Monoclonal Antibody [clone: 679340]

Supplier: R & D Systems
MAB5640
102886-066EA 633.42 USD
102886-066
Anti-NFATC1 (phospho-Ser172) Mouse Monoclonal Antibody [clone: 679340]
Antibodies
Type: Primary
Antigen Symbol: NFATC1
Antigen Name: nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Antigen Synonyms: NF-ATC, NF-ATc1.2, NFAT2, NFATc
Clonality: Monoclonal
Clone: 679340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR