Anti-LCP2 Mouse Monoclonal Antibody [clone: 460004]

Supplier: R & D Systems
MAB5637
220021-217EA 594.37 USD
220021-217
Anti-LCP2 Mouse Monoclonal Antibody [clone: 460004]
Antibodies
Type: Primary
Antigen: LCP2
Clonality: Monoclonal
Clone: 460004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR