Anti-TERF2 Mouse Monoclonal Antibody [clone: 689524]

Supplier: R & D Systems
MAB5635
102886-064EA 584.25 USD
102886-064
Anti-TERF2 Mouse Monoclonal Antibody [clone: 689524]
Antibodies
Type: Primary
Antigen: TERF2
Clonality: Monoclonal
Clone: 689524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR