Anti-ARNT Mouse Monoclonal Antibody [clone: 775146]

Supplier: R & D Systems
MAB5630
102886-062EA 577.02 USD
102886-062
Anti-ARNT Mouse Monoclonal Antibody [clone: 775146]
Antibodies
Type: Primary
Antigen: ARNT
Clonality: Monoclonal
Clone: 775146
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR