Anti-IL1R2 Rat Monoclonal Antibody [clone: 130817]

Supplier: R & D Systems
MAB563
220021-216EA 763.57 USD
220021-216
Anti-IL1R2 Rat Monoclonal Antibody [clone: 130817]
Antibodies
Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 130817
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR