Anti-NF2 Mouse Monoclonal Antibody [clone: 534428]

Supplier: R & D Systems
MAB5616
220021-212EA 594.37 USD
220021-212
Anti-NF2 Mouse Monoclonal Antibody [clone: 534428]
Antibodies
Type: Primary
Antigen Symbol: NF2
Antigen Name: neurofibromin 2 (merlin)
Antigen Synonyms: ACN, BANF, SCH
Clonality: Monoclonal
Clone: 534428
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR