Anti-SIGLEC1 Rat Monoclonal Antibody [clone: 645608]

Supplier: R & D Systems
MAB5610
102886-060EA 600.15 USD
102886-060
Anti-SIGLEC1 Rat Monoclonal Antibody [clone: 645608]
Antibodies
Type: Primary
Antigen: SIGLEC1
Clonality: Monoclonal
Clone: 645608
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR