Anti-GFRA1 Mouse Monoclonal Antibody [clone: 81401]

Supplier: R & D Systems
MAB560
220021-210EA 763.57 USD
220021-210
Anti-GFRA1 Mouse Monoclonal Antibody [clone: 81401]
Antibodies
Type: Primary
Antigen Symbol: GFRA1
Antigen Name: GDNF family receptor alpha 1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 81401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR