Anti-PPM1G Mouse Monoclonal Antibody [clone: 528322]

Supplier: R & D Systems
MAB5595
220021-209EA 577.02 USD
220021-209
Anti-PPM1G Mouse Monoclonal Antibody [clone: 528322]
Antibodies
Type: Primary
Antigen: PPM1G
Clonality: Monoclonal
Clone: 528322
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR