Anti-CAMK2A Mouse Monoclonal Antibody [clone: 525501]

Supplier: R & D Systems
MAB5584
220021-208EA 577.02 USD
220021-208
Anti-CAMK2A Mouse Monoclonal Antibody [clone: 525501]
Antibodies
Type: Primary
Antigen: CAMK2A
Clonality: Monoclonal
Clone: 525501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR