Anti-ADGRE1 Rat Monoclonal Antibody [clone: 52124]

Supplier: R & D Systems
MAB5580
220021-207EA 616.06 USD
220021-207
Anti-ADGRE1 Rat Monoclonal Antibody [clone: 52124]
Antibodies
Type: Primary
Antigen: ADGRE1
Clonality: Monoclonal
Clone: 52124
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR