Anti-EDN3 Mouse Monoclonal Antibody [clone: 3B6]

Supplier: R & D Systems
MAB5579
220021-206EA 577.02 USD
220021-206
Anti-EDN3 Mouse Monoclonal Antibody [clone: 3B6]
Antibodies
Type: Primary
Antigen: EDN3
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR