Anti-S100 A9 Mouse Monoclonal Antibody [clone: 474315]

Supplier: R & D Systems
MAB5578
220021-205EA 600.15 USD
220021-205
Anti-S100 A9 Mouse Monoclonal Antibody [clone: 474315]
Antibodies
Type: Primary
Antigen: S100 A9
Clonality: Monoclonal
Clone: 474315
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR