Anti-MESDC2 Mouse Monoclonal Antibody [clone: 513802]

Supplier: R & D Systems
MAB5577
102886-058EA 577.02 USD
102886-058
Anti-MESDC2 Mouse Monoclonal Antibody [clone: 513802]
Antibodies
Type: Primary
Antigen: MESDC2
Clonality: Monoclonal
Clone: 513802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR