Anti-TXLNA Mouse Monoclonal Antibody [clone: 812901]

Supplier: R & D Systems
MAB5575
102886-056EA 577.02 USD
102886-056
Anti-TXLNA Mouse Monoclonal Antibody [clone: 812901]
Antibodies
Type: Primary
Antigen: TXLNA
Clonality: Monoclonal
Clone: 812901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR