Anti-SEMA3C Mouse Monoclonal Antibody [clone: 757820]

Supplier: R & D Systems
MAB5570
102886-054EA 594.37 USD
102886-054
Anti-SEMA3C Mouse Monoclonal Antibody [clone: 757820]
Antibodies
Type: Primary
Antigen: SEMA3C
Clonality: Monoclonal
Clone: 757820
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR