Anti-CNTF Mouse Monoclonal Antibody [clone: 34738]

Supplier: R & D Systems
MAB557
220021-203EA 763.57 USD
220021-203
Anti-CNTF Mouse Monoclonal Antibody [clone: 34738]
Antibodies
Type: Primary
Antigen: CNTF
Clonality: Monoclonal
Clone: 34738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR