Anti-ADAM33 Mouse Monoclonal Antibody [clone: 5811]

Supplier: R & D Systems
MAB5565
220021-202EA 577.02 USD
220021-202
Anti-ADAM33 Mouse Monoclonal Antibody [clone: 5811]
Antibodies
Type: Primary
Antigen: ADAM33
Clonality: Monoclonal
Clone: 5811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR