Anti-CKM Mouse Monoclonal Antibody [clone: 492731]

Supplier: R & D Systems
MAB5564
220021-201EA 594.37 USD
220021-201
Anti-CKM Mouse Monoclonal Antibody [clone: 492731]
Antibodies
Type: Primary
Antigen: CKM
Clonality: Monoclonal
Clone: 492731
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR