Anti-NGF Mouse Monoclonal Antibody [clone: 884530]

Supplier: R & D Systems
MAB556
102886-052EA 577.02 USD
102886-052
Anti-NGF Mouse Monoclonal Antibody [clone: 884530]
Antibodies
Type: Primary
Antigen Symbol: NGF
Antigen Name: nerve growth factor (beta polypeptide)
Antigen Synonyms: Ngfb, beta-NGF
Clonality: Monoclonal
Clone: 884530
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR