Anti-SULT1A1 Mouse Monoclonal Antibody [clone: 638708]

Supplier: R & D Systems
MAB5546
102886-050EA 577.02 USD
102886-050
Anti-SULT1A1 Mouse Monoclonal Antibody [clone: 638708]
Antibodies
Type: Primary
Antigen: SULT1A1
Clonality: Monoclonal
Clone: 638708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR