Anti-SULT1E1 Mouse Monoclonal Antibody [clone: 726211]

Supplier: R & D Systems
MAB5545
102886-048EA 577.02 USD
102886-048
Anti-SULT1E1 Mouse Monoclonal Antibody [clone: 726211]
Antibodies
Type: Primary
Antigen: SULT1E1
Clonality: Monoclonal
Clone: 726211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR