Anti-CD4 Rat Monoclonal Antibody [clone: 131922]

Supplier: R & D Systems
MAB5541
220021-198EA 763.57 USD
220021-198
Anti-CD4 Rat Monoclonal Antibody [clone: 131922]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 131922
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR