Anti-GPER1 Mouse Monoclonal Antibody [clone: 730737]

Supplier: R & D Systems
MAB5534
102886-046EA 577.02 USD
102886-046
Anti-GPER1 Mouse Monoclonal Antibody [clone: 730737]
Antibodies
Type: Primary
Antigen Symbol: GPER1
Antigen Name: G protein-coupled estrogen receptor 1
Antigen Synonyms: CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
Clonality: Monoclonal
Clone: 730737
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR