Anti-SNCB Mouse Monoclonal Antibody [clone: 516505]

Supplier: R & D Systems
MAB5528
220021-196EA 594.37 USD
220021-196
Anti-SNCB Mouse Monoclonal Antibody [clone: 516505]
Antibodies
Type: Primary
Antigen: SNCB
Clonality: Monoclonal
Clone: 516505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR