Anti-JUND Mouse Monoclonal Antibody [clone: 512709]

Supplier: R & D Systems
MAB5526
220021-195EA 594.37 USD
220021-195
Anti-JUND Mouse Monoclonal Antibody [clone: 512709]
Antibodies
Type: Primary
Antigen: JUND
Clonality: Monoclonal
Clone: 512709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR