Anti-IL1A Mouse Monoclonal Antibody [clone: 140221]

Supplier: R & D Systems
MAB552
220021-193EA 763.57 USD
220021-193
Anti-IL1A Mouse Monoclonal Antibody [clone: 140221]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 140221
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR