Anti-CCRL2 Rat Monoclonal Antibody [clone: 498321]

Supplier: R & D Systems
MAB5519
102886-044EA 594.37 USD
102886-044
Anti-CCRL2 Rat Monoclonal Antibody [clone: 498321]
Antibodies
Type: Primary
Antigen: CCRL2
Clonality: Monoclonal
Clone: 498321
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR