Anti-IL15RA Rat Monoclonal Antibody [clone: 888220]

Supplier: R & D Systems
MAB5511
102886-042EA 577.02 USD
102886-042
Anti-IL15RA Rat Monoclonal Antibody [clone: 888220]
Antibodies
Type: Primary
Antigen: IL15RA
Clonality: Monoclonal
Clone: 888220
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR