Anti-IL15RA Rat Monoclonal Antibody [clone: 146018]

Supplier: R & D Systems
MAB551
220021-192EA 763.57 USD
220021-192
Anti-IL15RA Rat Monoclonal Antibody [clone: 146018]
Antibodies
Type: Primary
Antigen: IL15RA
Clonality: Monoclonal
Clone: 146018
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR