Anti-HMGB3 Mouse Monoclonal Antibody [clone: 546519]

Supplier: R & D Systems
MAB55071
102886-040EA 577.02 USD
102886-040
Anti-HMGB3 Mouse Monoclonal Antibody [clone: 546519]
Antibodies
Type: Primary
Antigen: HMGB3
Clonality: Monoclonal
Clone: 546519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR