Anti-HMGB3 Mouse Monoclonal Antibody [clone: 546737]

Supplier: R & D Systems
MAB5507
220021-191EA 577.02 USD
220021-191
Anti-HMGB3 Mouse Monoclonal Antibody [clone: 546737]
Antibodies
Type: Primary
Antigen: HMGB3
Clonality: Monoclonal
Clone: 546737
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR