Anti-TREML4 Rat Monoclonal Antibody [clone: 817914]

Supplier: R & D Systems
MAB5500
102886-038EA 584.25 USD
102886-038
Anti-TREML4 Rat Monoclonal Antibody [clone: 817914]
Antibodies
Type: Primary
Antigen: TREML4
Clonality: Monoclonal
Clone: 817914
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR