Anti-AGRN Mouse Monoclonal Antibody [clone: 7685]

Supplier: R & D Systems
MAB550
220021-190EA 763.57 USD
220021-190
Anti-AGRN Mouse Monoclonal Antibody [clone: 7685]
Antibodies
Type: Primary
Antigen: AGRN
Clonality: Monoclonal
Clone: 7685
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR