Anti-CSF2RB2 Rat Monoclonal Antibody [clone: 130710]

Supplier: R & D Systems
MAB5491
220021-188EA 763.57 USD
220021-188
Anti-CSF2RB2 Rat Monoclonal Antibody [clone: 130710]
Antibodies
Type: Primary
Antigen: CSF2RB2
Clonality: Monoclonal
Clone: 130710
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR