Anti-CSF2RB2 Rat Monoclonal Antibody [clone: 130714]

Supplier: R & D Systems
MAB549
220021-187EA 763.57 USD
220021-187
Anti-CSF2RB2 Rat Monoclonal Antibody [clone: 130714]
Antibodies
Type: Primary
Antigen: CSF2RB2
Clonality: Monoclonal
Clone: 130714
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR