Anti-NELL1 Mouse Monoclonal Antibody [clone: 756435]

Supplier: R & D Systems
MAB54871
102886-036EA 600.15 USD
102886-036
Anti-NELL1 Mouse Monoclonal Antibody [clone: 756435]
Antibodies
Type: Primary
Antigen Symbol: NELL1
Antigen Name: neural EGFL like 1
Antigen Synonyms: IDH3GL, NRP1
Clonality: Monoclonal
Clone: 756435
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR