Anti-NELL1 Mouse Monoclonal Antibody [clone: 756465]

Supplier: R & D Systems
MAB5487
102886-034EA 577.02 USD
102886-034
Anti-NELL1 Mouse Monoclonal Antibody [clone: 756465]
Antibodies
Type: Primary
Antigen Symbol: NELL1
Antigen Name: neural EGFL like 1
Antigen Synonyms: IDH3GL, NRP1
Clonality: Monoclonal
Clone: 756465
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR