Anti-PLXNA2 Rat Monoclonal Antibody [clone: 583603]

Supplier: R & D Systems
MAB5486
102886-032EA 577.02 USD
102886-032
Anti-PLXNA2 Rat Monoclonal Antibody [clone: 583603]
Antibodies
Type: Primary
Antigen: PLXNA2
Clonality: Monoclonal
Clone: 583603
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR