Anti-SEMA4B Mouse Monoclonal Antibody [clone: 561416]

Supplier: R & D Systems
MAB5485
220021-186EA 600.15 USD
220021-186
Anti-SEMA4B Mouse Monoclonal Antibody [clone: 561416]
Antibodies
Type: Primary
Antigen: SEMA4B
Clonality: Monoclonal
Clone: 561416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR