Anti-CFHR2 Mouse Monoclonal Antibody [clone: 510511]

Supplier: R & D Systems
MAB5484
220021-185EA 594.37 USD
220021-185
Anti-CFHR2 Mouse Monoclonal Antibody [clone: 510511]
Antibodies
Type: Primary
Antigen: CFHR2
Clonality: Monoclonal
Clone: 510511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR