Anti-MMP3 Rat Monoclonal Antibody [clone: 145106]

Supplier: R & D Systems
MAB548
220021-183EA 772.25 USD
220021-183
Anti-MMP3 Rat Monoclonal Antibody [clone: 145106]
Antibodies
Type: Primary
Antigen: MMP3
Clonality: Monoclonal
Clone: 145106
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR