Anti-GPX2 Mouse Monoclonal Antibody [clone: 496010]

Supplier: R & D Systems
MAB5470
220021-181EA 594.37 USD
220021-181
Anti-GPX2 Mouse Monoclonal Antibody [clone: 496010]
Antibodies
Type: Primary
Antigen Symbol: GPX2
Antigen Name: glutathione peroxidase 2
Antigen Synonyms: GI-GPx, GPRP, GPRP-2, GPx-2, GPx-GI, GSHPX-GI, GSHPx-2
Clonality: Monoclonal
Clone: 496010
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR