Anti-MGAT5 Mouse Monoclonal Antibody [clone: 706824]

Supplier: R & D Systems
MAB5469
102886-030EA 600.15 USD
102886-030
Anti-MGAT5 Mouse Monoclonal Antibody [clone: 706824]
Antibodies
Type: Primary
Antigen: MGAT5
Clonality: Monoclonal
Clone: 706824
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR