Anti-KLF2 Mouse Monoclonal Antibody [clone: 665333]

Supplier: R & D Systems
MAB5466
102886-028EA 594.37 USD
102886-028
Anti-KLF2 Mouse Monoclonal Antibody [clone: 665333]
Antibodies
Type: Primary
Antigen: KLF2
Clonality: Monoclonal
Clone: 665333
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR